Merck
  • Thymosin beta 4 attenuates PrP(106-126)-induced human brain endothelial cells dysfunction.
Thymosin beta 4 attenuates PrP(106-126)-induced human brain endothelial cells dysfunction.
European journal of pharmacology (2019-12-27)
Kibbeum Song, Hye-Ju Han, Sokho Kim, Jungkee Kwon
ABSTRACT
The blood-brain barrier (BBB) is a highly selective permeability barrier that separates the circulating blood from the brain and extracellular fluid in the central nervous system (CNS). The BBB is formed by cerebral endothelial cells connected by tight junctions. Prion diseases are neurodegenerative pathologies characterized by the accumulation of altered forms of the prion protein (PrP), named PrPSc. Thymosin beta 4 (Tβ4) is an actin-sequestering peptide known to bind monomeric actin and inhibit its polymerization, and it is known to have a neuroprotective effect. However, the effect of Tβ4 on prion disease has not yet been investigated. Therefore, in this study, we investigated the effect of Tβ4 on prion-induced BBB dysfunction in hCMEC/D3 human cerebral endothelial cells. We found that Tβ4 increased the expression of tight junction protein, but reduced the ratio of F-actin to G-actin. Moreover, we showed that Tβ4 significantly improved PrP (106-126)-induced vascular permeability dysfunction in hCMEC/D3 cells. Through human BBB in vitro model, we found that PrP (106-126) could disrupt tight junctions and cytoskeleton arrangement. These results suggest that Tβ4 may play a critical role in barrier stabilization. Furthermore, Tβ4 may prevent neurodegenerative diseases caused by prion-induced BBB dysfunction.
Sigma-Aldrich
In Vitro Vascular Permeability Assay (96-well), This In Vitro Vascular Permeability Assay kit employs a 96-well plate, and provides an efficient system for evaluating the effects of chemicals & drug compounds on endothelial cell adsorption, transport & permeability.