Merck
  • Development and validation of LC-ESI-MS/MS methods for quantification of 27 free and conjugated estrogen-related metabolites.
Development and validation of LC-ESI-MS/MS methods for quantification of 27 free and conjugated estrogen-related metabolites.
Analytical biochemistry (2019-12-06)
Carien van der Berg, Gerda Venter, Francois H van der Westhuizen, Elardus Erasmus
ABSTRACT
An imbalance in the estrogen metabolism has been associated with an increased risk of breast cancer development. Evaluation of the estrogen biotransformation capacity requires monitoring of various estrogen metabolites. Up to now, only some estrogen metabolites could be measured in urine. However, in order to offer tailor made nutritional support or therapies, a complete estrogen metabolite profile is required in order to identify specific deficiencies in this pathway for each patient individually. Here, we focused on this need to quantify as many as possible of the estrogen-related metabolites excreted in urine. The method was developed to quantify 27 estrogen-related metabolites in small urine quantities. This entailed sample clean-up with a multi-step solid phase extraction procedure, derivatisation of the metabolites in the less water-soluble fraction through dansylation, and analyses using liquid chromatography-tandem mass spectrometry (LC-MS/MS). The metabolites accurately quantified by the method devised included parent estrogens, hydroxylated and methylated forms, metabolites of the 16α-hydroxyestrogen pathway, sulphate and glucuronide conjugated forms, precursors and a related steroid hormone. This method was validated and enabled quantification in the high picograms and low nanograms per millilitre range. Finally, analyses of urine samples confirmed detection and quantification of each of the metabolites.
Product Description
Supelco
4-Androstene-3,17-dione, VETRANAL®, analytical standard
Sigma-Aldrich
2-Acetamidophenol, 97%