Merck
  • Expression of the RNA methyltransferase Nsun5 is essential for developing cerebral cortex.
Expression of the RNA methyltransferase Nsun5 is essential for developing cerebral cortex.
Molecular brain (2019-08-30)
Peipei Chen, Tingting Zhang, Zihao Yuan, Bin Shen, Ling Chen
ABSTRACT
Nsun5 gene, encoding a cytosine-5 RNA methyltransferase, is deleted in about 95% patients with Williams-Beuren syndrome (WBS). WBS is a neurodevelopmental disorder and characterized by cognitive disorder. We generated single-gene Nsun5 knockout (Nsun5-KO) mice and reported that the Nsun5 deletion leads to deficit in spatial cognition. This study focused on investigating the influence of Nsun5 deficiency in the development of cerebral cortex. In comparison with wild-type littermates, the cortical thickness in postnatal day 10 Nsun5-KO mice was obviously reduced with an abnormal laminar organization, and the processes of pyramidal cells were shorter and finer. Nsun5 was selectively expressed in radial glial cells (RGCs) of cerebral cortex from embryonic day (E) 12.5 to E16.5, but not in intermediate progenitor cells (IPCs) or neocortical neurons. The Nsun5 deletion did not alter proliferation of RGCs or differentiation of RGCs into IPCs. Notably, the ablation of Nsun5 disrupted the growth of radial glial scaffolds, thus numerous basal processes of RGCs failed to reach pial basement membrane. Level of cell polarity regulator Cdc42 protein in radial glial scaffolds of E14.5 Nsun5-KO mice was reduced, but the level of Cdc42 mRNA was unchanged. The dysfunction of glial scaffolds impeded the radial migration of upper-layer and deeper-layer neurons to cause their subcortical accumulation and apoptosis, resulting in an obvious thinness of the cortical plate in E18.5 Nsun5-KO mice. These findings establish a critical role of Nsun5 in development of cerebral cortex through regulating radial glial scaffolds of RGCs to control migration of neocortical neurons.
Sigma-Aldrich
Anti-BrdU Antibody, clone 131-14871, clone 131-14871, Chemicon®, from mouse