Merck
Nucleocytoplasmic shuttling of endocytic proteins.
The Journal of cell biology (2001-06-27)
M Vecchi, S Polo, V Poupon, J W van de Loo, A Benmerah, P P Di Fiore
ABSTRACT
Many cellular processes rely on the ordered assembly of macromolecular structures. Here, we uncover an unexpected link between two such processes, endocytosis and transcription. Many endocytic proteins, including eps15, epsin1, the clathrin assembly lymphoid myeloid leukemia (CALM), and alpha-adaptin, accumulate in the nucleus when nuclear export is inhibited. Endocytosis and nucleocytoplasmic shuttling of endocytic proteins are apparently independent processes, since inhibition of endocytosis did not appreciably alter nuclear translocation of endocytic proteins, and blockade of nuclear export did not change the initial rate of endocytosis. In the nucleus, eps15 and CALM acted as positive modulators of transcription in a GAL4-based transactivation assay, thus raising the intriguing possibility that some endocytic proteins play a direct or indirect role in transcriptional regulation.
Sigma-Aldrich
Monoclonal Anti-β1 and β2-Adaptins antibody produced in mouse, clone 100/1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-γ-Adaptin antibody produced in mouse, clone 100/3, ascites fluid