Merck
  • Ectopic expression of transcription factor AP-2δ in developing retina: effect on PSA-NCAM and axon routing.
Ectopic expression of transcription factor AP-2δ in developing retina: effect on PSA-NCAM and axon routing.
Journal of neurochemistry (2013-11-06)
Xiaodong Li, Elizabeth A Monckton, Roseline Godbout
ABSTRACT
Retinal ganglion cells transmit the visual signal from the retina to the brain. We have previously shown that the activator protein 2 (AP-2)δ (TFAP2D) transcription factor is expressed in one third of ganglion cells in developing retina suggesting a specialized role for these AP-2δ-expressing cells. Here, we address the role of AP-2δ in retina by in ovo electroporation of RCAS/AP-2δ retroviral constructs into the eyes of chick embryos at day 2 of gestation. Ectopic expression of AP-2δ does not affect lineage differentiation in the developing retina. However, immunostaining of retinal tissue with markers associated with axonal growth such as growth-associated protein 43 and polysialic acid-neural cell adhesion molecule (PSA-NCAM) demonstrates axonal misrouting and abnormal axonal bundling. Treatment of AP-2δ-misexpressing retinal cell cultures with endoneuraminidase, an enzyme that removes PSA from NCAM, decreases AP-2δ-induced axonal bundling. Our data suggest a role for AP-2δ in polysialylation of NCAM, with ectopic expression of AP-2δ resulting in premature bundling of emerging axons and misrouting of axons. We propose that expression of AP-2δ in a subset of ganglion cells contributes to the fine-tuning of axonal growth in the developing retina.
Sigma-Aldrich
Monoclonal Anti-Growth Associated Protein-43 antibody produced in mouse, clone GAP-7B10, ascites fluid