Merck
  • Identification of TBK1 complexes required for the phosphorylation of IRF3 and the production of interferon β.
Identification of TBK1 complexes required for the phosphorylation of IRF3 and the production of interferon β.
The Biochemical journal (2017-02-06)
Siddharth Bakshi, Jordan Taylor, Sam Strickson, Thomas McCartney, Philip Cohen
ABSTRACT
The double-stranded RNA mimetic poly(I:C) and lipopolysaccharide (LPS) activate Toll-like receptors 3 (TLR3) and TLR4, respectively, triggering the activation of TANK (TRAF family member-associated NF-κB activator)-binding kinase 1 (TBK1) complexes, the phosphorylation of interferon regulatory factor 3 (IRF3) and transcription of the interferon β (IFNβ) gene. Here, we demonstrate that the TANK-TBK1 and optineurin (OPTN)-TBK1 complexes control this pathway. The poly(I:C)- or LPS-stimulated phosphorylation of IRF3 at Ser396 and production of IFNβ were greatly reduced in bone marrow-derived macrophages (BMDMs) from TANK knockout (KO) mice crossed to knockin mice expressing the ubiquitin-binding-defective OPTN[D477N] mutant. In contrast, IRF3 phosphorylation and IFNβ production were not reduced significantly in BMDM from OPTN[D477N] knockin mice and only reduced partially in TANK KO BMDM. The TLR3/TLR4-dependent phosphorylation of IRF3 and IFNβ gene transcription were not decreased in macrophages from OPTN[D477N] crossed to mice deficient in IκB kinase ε, a TANK-binding kinase related to TBK1. In contrast with the OPTN-TBK1 complex, TBK1 associated with OPTN[D477N] did not undergo phosphorylation at Ser172 in response to poly(I:C) or LPS, indicating that the interaction of ubiquitin chains with OPTN is required to activate OPTN-TBK1 in BMDM. The phosphorylation of IRF3 and IFNβ production induced by Sendai virus infection were unimpaired in BMDM from TANK KO × OPTN[D477N] mice, suggesting that other/additional TBK1 complexes control the RIG-I-like receptor-dependent production of IFNβ. Finally, we present evidence that, in human HACAT cells, the poly(I:C)-dependent phosphorylation of TBK1 at Ser172 involves a novel TBK1-activating kinase(s).
Sigma-Aldrich
Anti-IKKβ Antibody, clone 10AG2, clone 10AG2, Upstate®, from mouse