Merck
  • A quantitative analytical method to test for salt effects on giant unilamellar vesicles.
A quantitative analytical method to test for salt effects on giant unilamellar vesicles.
Scientific reports (2012-02-23)
Maik Hadorn, Eva Boenzli, Peter Eggenberger Hotz
ABSTRACT
Today, free-standing membranes, i.e. liposomes and vesicles, are used in a multitude of applications, e.g. as drug delivery devices and artificial cell models. Because current laboratory techniques do not allow handling of large sample sizes, systematic and quantitative studies on the impact of different effectors, e.g. electrolytes, are limited. In this work, we evaluated the Hofmeister effects of ten alkali metal halides on giant unilamellar vesicles made of palmitoyloleoylphosphatidylcholine for a large sample size by combining the highly parallel water-in-oil emulsion transfer vesicle preparation method with automatic haemocytometry. We found that this new quantitative screening method is highly reliable and consistent with previously reported results. Thus, this method may provide a significant methodological advance in analysis of effects on free-standing model membranes.
Sigma-Aldrich
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, ≥95.5% (GC), ≥98% (TLC)
Sigma-Aldrich
Mineral oil, light