Merck
Single-cell transcriptomic profiling of the aging mouse brain.
Nature neuroscience (2019-09-26)
Methodios Ximerakis, Scott L Lipnick, Brendan T Innes, Sean K Simmons, Xian Adiconis, Danielle Dionne, Brittany A Mayweather, Lan Nguyen, Zachary Niziolek, Ceren Ozek, Vincent L Butty, Ruth Isserlin, Sean M Buchanan, Stuart S Levine, Aviv Regev, Gary D Bader, Joshua Z Levin, Lee L Rubin
ABSTRACT
The mammalian brain is complex, with multiple cell types performing a variety of diverse functions, but exactly how each cell type is affected in aging remains largely unknown. Here we performed a single-cell transcriptomic analysis of young and old mouse brains. We provide comprehensive datasets of aging-related genes, pathways and ligand-receptor interactions in nearly all brain cell types. Our analysis identified gene signatures that vary in a coordinated manner across cell types and gene sets that are regulated in a cell-type specific manner, even at times in opposite directions. These data reveal that aging, rather than inducing a universal program, drives a distinct transcriptional course in each cell population, and they highlight key molecular processes, including ribosome biogenesis, underlying brain aging. Overall, these large-scale datasets (accessible online at https://portals.broadinstitute.org/single_cell/study/aging-mouse-brain ) provide a resource for the neuroscience community that will facilitate additional discoveries directed towards understanding and modifying the aging process.
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse
Sigma-Aldrich
D-(+)-Trehalose dihydrate, from Saccharomyces cerevisiae, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Calcein Blue, fluorescent dye