Merck
  • LncRNA-CTS promotes metastasis and epithelial-to-mesenchymal transition through regulating miR-505/ZEB2 axis in cervical cancer.
LncRNA-CTS promotes metastasis and epithelial-to-mesenchymal transition through regulating miR-505/ZEB2 axis in cervical cancer.
Cancer letters (2019-09-10)
Shujun Feng, Wei Liu, Xiaoxu Bai, Wenjing Pan, Zhaoyang Jia, Shunjin Zhang, Yimin Zhu, Wenhua Tan
ABSTRACT
Cervical carcinoma (CC) is the second most common cancer in females. In order to improve current anti-metastasis strategies for CC, it is important to improve our understanding of the mechanisms involved in epithelial-to-mesenchymal transition (EMT). This study aimed to elucidate the potential role of a novel long non-coding RNA (lncRNA)-CTS and the mechanisms underlying EMT in CC. The expression levels of lncRNA-CTS and miR-505 were detected using quantitative reverse transcriptase polymerase chain reaction in CC specimens and cells (HeLa, SiHa, Ca-Ski, C-33A, and HT-3). Further experiments including wound scratch and transwell invasion assays, Western blotting, immunofluorescence, and luciferase assays were used to investigate the function of lncRNA-CTS/miR-505/ZEB2 in vitro. In addition, a tumor xenograft model was used to assess the effect of lncRNA-CTS in vivo. The expression levels of lncRNA-CTS and miR-505 were correlated with the metastasis-associated clinicopathological features of CC patients. Moreover, lncRNA-CTS was associated with a poor prognosis in CC patients. In vitro and in vivo experiments, along with gain- and loss-of-function studies, showed that lncRNA-CTS enhanced cell migration, invasion, and the transforming growth factor (TGF)-β1-induced-EMT process. Data also showed that lncRNA-CTS could function as a competing endogenous RNA for miR-505 in CC cells. Further investigations disclosed that ZEB2 was demonstrated as a downstream target of miR-505, and subsequently exerted its metastatic effects via the lncRNA-CTS/miR-505/ZEB2 axis in CC cells. Finally, lncRNA-CTS activated the SMAD/TGF pathway via miR-505 in CC cells. Collectively, our results demonstrate the importance of the lncRNA-CTS/miR-505/ZEB2 axis in CC. LncRNA-CTS can predispose CC patients to metastases and may represent a promising therapeutic target for CC.
Sigma-Aldrich
MISSION® esiRNA, targeting human TTR