Merck
Reduction Expansion Synthesis of Sintered Metal.
Materials (Basel, Switzerland) (2019-09-11)
Zachary Daniels, Wilson Rydalch, Troy Y Ansell, Claudia C Luhrs, Jonathan Phillips
ABSTRACT
Described herein is a novel method, Reduction Expansion Synthesis-Sintered Metal (RES-SM), to create a sintered metal body of a designed shape at ambient pressure, hundreds of degrees below the metal melting temperature. The precursor to the metal part is a mixture of metal oxide particles and activated metal particles, and in this study specifically nickel oxide and activated nickel metal particles. It is postulated that the metal oxide component is reduced via exposure to chemical radical species produced via thermal decomposition of urea or other organic compounds. In the study performed, the highest temperature required was 950 °C, the longest duration of high temperature treatment was 1200 s, and in all cases, the atmosphere was inert gas at ambient pressure. As discovered using scanning electron microscopy (SEM), energy dispersive spectroscopy (EDS) and x-ray diffraction (XRD), the metal that forms via the RES process presents necks of completely reduced metal between existing metal particles. The 'as produced' parts are similar in properties to 'brown' metal parts created using more standard methods and require 'post processing' to full densify. Parts treated by hot isostatic pressing show fully self-supporting, robust structures, with hardness values like others reported in literature for traditional fabrication methods. This novel method uses affordable and environmentally friendly precursors to join metallic parts at moderate temperatures, produces fully reduced metals in a very short time and has potential to make many parts simultaneously in a standard laboratory furnace.
Sigma-Aldrich
Nickel (IV) oxide, technical, oxidizing agent, ~30% active peroxide basis