Merck
  • Anti-Hypochlorite and Catalytic Activity of Commercially Available Moringa oleifera Diet Supplement.
Anti-Hypochlorite and Catalytic Activity of Commercially Available Moringa oleifera Diet Supplement.
Molecules (Basel, Switzerland) (2019-09-25)
Karolina Starzak, Bernadette Creaven, Arkadiusz Matwijczuk, Alicja Matwijczuk, Dariusz Karcz
ABSTRACT
Aiming at the assessment of the pro-health, and especially anti-hypochlorite properties of Moringa oleifera species a representative, commercially available Moringa oleifera dietary supplement was used as a substrate for the preparation of aqueous Moringa extract. The anti-hypochlorite activity of the extract was assessed using the hypochlorite-specific coumarin-based fluorescence turn-off sensor, namely 7-diethylamino-coumarin-3-carboxylic acid (7-DCCA). This compound was synthesized via the Knoevenagel condensation of 4-diethylamino-2-hydroxybenzaldehyde with Meldrum's acid and the Moringa extract was employed as a medium and catalyst. Moreover, the total phenolic content (TPC) as well as the reactive oxygen species (ROS)-scavenging ability of the aqueous Moringa extract were determined. The results obtained demonstrated the applicability of Moringa extract as an anti-hypochlorite agent. Additionally, the satisfactory yield of the 7-DCCA obtained suggests the usefulness of the extract as a catalyst and the reaction medium. The antioxidative potential of the extract was notably lower than that of the standard (TROLOX). Determination of TPC in 100 g of the dry weight (DW) of studied material revealed a high number of polyphones present.
Sigma-Aldrich
4-(Diethylamino)salicylaldehyde, 98%
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%