Merck
  • Comparison of covalent with reversible inhibitor binding sites of the gastric H,K-ATPase by site-directed mutagenesis.
Comparison of covalent with reversible inhibitor binding sites of the gastric H,K-ATPase by site-directed mutagenesis.
The Journal of biological chemistry (2000-02-08)
N Lambrecht, K Munson, O Vagin, G Sachs
ABSTRACT
The gastric H,K-ATPase is covalently inhibited by substituted pyridyl-methylsulfinyl-benzimidazoles, such as omeprazole, that convert to thiophilic probes of luminally accessible cysteines in the acid space. The K(+) competitive inhibitor, SCH28080, prevented inhibition of acid transport by omeprazole. In stably expressing HEK293 cells, the benzimidazole-reactive cysteines, Cys-321 (transmembrane helix (TM) 3), Cys-813 and Cys-822 (TM5/6), and Cys-892 (TM7/8) were mutated to the amino acids found in the SCH28080-resistant Na,K-ATPase and kinetic parameters of H,K-ATPase activity analyzed. Mutations of Cys-822 and Cys-892 had insignificant effects on the K(i(app)), K(m(app)) or V(max), but mutations of Cys-813 to threonine and Cys-321 to alanine decreased the affinity for SCH28080. Mutation of Cys-321 to alanine produced mixed kinetics of inhibition, still with higher affinity for the cation-free form of phosphoenzyme. Since the phenylmethoxy ring of the imidazo-pyridine inhibitors binds to TM1/2, as shown by earlier photoaffinity studies, and the mutations in TM6 (Cys-813 --> Thr) as well as the end of TM3 (Cys-321 --> Ala) decrease the affinity for SCH28080, the TM1/2, TM3, and TM6 helices lie within approximately 16 A of each other based on the size of the active, extended conformation of SCH28080.
Sigma-Aldrich
Omeprazole, A cell-permeable pyridyl methylsulfinyl benzimidazole compound that acts as selective proton pump inhibitor.