Merck
  • Molecular mechanism and potential target indication of TAK-931, a novel CDC7-selective inhibitor.
Molecular mechanism and potential target indication of TAK-931, a novel CDC7-selective inhibitor.
Science advances (2019-05-28)
Kenichi Iwai, Tadahiro Nambu, Ryo Dairiki, Momoko Ohori, Jie Yu, Kristine Burke, Masamitsu Gotou, Yukiko Yamamoto, Shunsuke Ebara, Sachio Shibata, Ryosuke Hibino, Satoru Nishizawa, Tohru Miyazaki, Misaki Homma, Yuya Oguro, Takashi Imada, Nobuo Cho, Noriko Uchiyama, Akifumi Kogame, Toshiyuki Takeuchi, Osamu Kurasawa, Kazunori Yamanaka, Huifeng Niu, Akihiro Ohashi
ABSTRACT
Replication stress (RS) is a cancer hallmark; chemotherapeutic drugs targeting RS are widely used as treatments for various cancers. To develop next-generation RS-inducing anticancer drugs, cell division cycle 7 (CDC7) has recently attracted attention as a target. We have developed an oral CDC7-selective inhibitor, TAK-931, as a candidate clinical anticancer drug. TAK-931 induced S phase delay and RS. TAK-931-induced RS caused mitotic aberrations through centrosome dysregulation and chromosome missegregation, resulting in irreversible antiproliferative effects in cancer cells. TAK-931 exhibited significant antiproliferative activity in preclinical animal models. Furthermore, in indication-seeking studies using large-scale cell panel data, TAK-931 exhibited higher antiproliferative activities in RAS-mutant versus RAS-wild-type cells; this finding was confirmed in pancreatic patient-derived xenografts. Comparison analysis of cell panel data also demonstrated a unique efficacy spectrum for TAK-931 compared with currently used chemotherapeutic drugs. Our findings help to elucidate the molecular mechanisms for TAK-931 and identify potential target indications.