Merck
  • Pharmacology in vivo of the phenylindan derivative, Lu 19-005, a new potent inhibitor of dopamine, noradrenaline and 5-hydroxytryptamine uptake in rat brain.
Pharmacology in vivo of the phenylindan derivative, Lu 19-005, a new potent inhibitor of dopamine, noradrenaline and 5-hydroxytryptamine uptake in rat brain.
Naunyn-Schmiedeberg's archives of pharmacology (1985-04-01)
J Arnt, A V Christensen, J Hyttel
ABSTRACT
Behavioural effects on dopaminergic transmission of a phenylindane derivative, Lu 19-005 [(+/-)-trans-3-(3,4-dichlorophenyl)-N-methyl-l-indanamine, HCI], with potent inhibitory effect on dopamine (DA), noradrenaline (NA) and serotonin (5-HT) uptake in rats and the effect on DA, NA and 5-HT activity in mice have been studied and compared with those of other known DA, NA and 5-HT uptake inhibitors with different selectivity ratios. Lu 19-005 induced stereotyped behaviour after parenteral and oral administration with a duration of action of more than 24 h. The stereotyped licking and biting induced by Lu 19-005 was antagonized by reserpine and cis(Z)-flupentixol, but not affected by prazosin, p-chlorophenylalanine and alpha-methyl-p-tyrosine pretreatments. Metergoline slightly facilitated the onset of stereotypy. Lower doses of Lu 19-005 induced ipsilateral circling in unilaterally 6-hydroxy-DA-lesioned rats. Finally, Lu 19-005 antagonized the catalepsy induced by perphenazine. In mice, Lu 19-005 potentiated the apomorphine-induced gnawing, reversed tetrabenazine-induced ptosis and potentiated the behavioural effects of 5-HTP within a similar dose range. The effects of Lu 19-005 were compared with those of other reference compounds. Nomifensine had qualitatively similar effects in rats although of much shorter duration. In mice, nomifensine selectively reversed tetrabenazine-induced ptosis. Weaker effects in all test models were found with bupropion, LR 5182 and GBR 13.069, compounds with inhibitory effect on DA and NA uptake. The DA-, NA- and 5-HT-uptake inhibitor diclofensine, however, had no effect in rats except in the 6-hydroxy-DA-circling test and had low potency in mice. The specific 5-HT- and NA-uptake inhibitors citalopram and talsupram, respectively, were ineffective in all rat models.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
Indatraline hydrochloride, solid