Merck
  • Selective dysfunction of the vagal component of the baroreflex following cerebral ischemia: protection by ifenprodil and flunarizine.
Selective dysfunction of the vagal component of the baroreflex following cerebral ischemia: protection by ifenprodil and flunarizine.
European journal of pharmacology (1990-11-06)
J Kurihara, T Sahara, N Oda, H Tomita, H Kato
ABSTRACT
Baroreflex sensitivity assessed from the phenylephrine-induced reflex bradycardia was significantly decreased following 5 min global incomplete cerebral ischemia in pentobarbitalized dogs. Although bilateral vagotomy in the cervical region decreased baroreflex sensitivity by about 50% in sham-operated animals, it hardly affected the baroreflex in animals subjected to ischemia. The extent of the decrease in the influence of vagotomy on the baroreflex was dependent on the severity of ischemia in the dorsal medulla oblongata. In animals vagotomized before ischemia, no significant decrease in baroreflex sensitivity was observed following ischemia. Pretreatment with ifenprodil or flunarizine, 1 mg/kg i.v., 5 min prior to ischemia prevented the post-ischemic decrease in baroreflex sensitivity. Vagotomy decreased baroreflex sensitivity during the reperfusion period in these treated animals. These results suggest that the post-ischemic attenuation of reflex bradycardia may be due to a selective dysfunction of the vagal component of baroreflex, which can be prevented by the cerebroprotective agents.
Sigma-Aldrich
Ifenprodil (+)-tartrate salt