Merck
  • Manipulation of microbiota reveals altered callosal myelination and white matter plasticity in a model of Huntington disease.
Manipulation of microbiota reveals altered callosal myelination and white matter plasticity in a model of Huntington disease.
Neurobiology of disease (2019-02-26)
Carola I Radulescu, Marta Garcia-Miralles, Harwin Sidik, Costanza Ferrari Bardile, Nur Amirah Binte Mohammad Yusof, Hae Ung Lee, Eliza Xin Pei Ho, Collins Wenhan Chu, Emma Layton, Donovan Low, Paola Florez De Sessions, Sven Pettersson, Florent Ginhoux, Mahmoud A Pouladi
ABSTRACT
Structural and molecular myelination deficits represent early pathological features of Huntington disease (HD). Recent evidence from germ-free (GF) animals suggests a role for microbiota-gut-brain bidirectional communication in the regulation of myelination. In this study, we aimed to investigate the impact of microbiota on myelin plasticity and oligodendroglial population dynamics in the mixed-sex BACHD mouse model of HD. Ultrastructural analysis of myelin in the corpus callosum revealed alterations of myelin thickness in BACHD GF compared to specific-pathogen free (SPF) mice, whereas no differences were observed between wild-type (WT) groups. In contrast, myelin compaction was altered in all groups when compared to WT SPF animals. Levels of myelin-related proteins were generally reduced, and the number of mature oligodendrocytes was decreased in the prefrontal cortex under GF compared to SPF conditions, regardless of genotype. Minor differences in commensal bacteria at the family and genera levels were found in the gut microbiota of BACHD and WT animals housed in standard living conditions. Our findings indicate complex effects of a germ-free status on myelin-related characteristics, and highlight the adaptive properties of myelination as a result of environmental manipulation.
Sigma-Aldrich
Anti-Calnexin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®
Sigma-Aldrich
Anti-Ermin Antibody, clone 160, clone 160, from mouse