Merck
  • Elevated WBP2 Expression in HER2-positive Breast Cancers Correlates with Sensitivity to Trastuzumab-based Neoadjuvant Therapy: A Retrospective and Multicentric Study.
Elevated WBP2 Expression in HER2-positive Breast Cancers Correlates with Sensitivity to Trastuzumab-based Neoadjuvant Therapy: A Retrospective and Multicentric Study.
Clinical cancer research : an official journal of the American Association for Cancer Research (2018-12-30)
Shin-Ae Kang, Jye Swei Guan, Hock Jin Tan, Tinghine Chu, Aye Aye Thike, Cristina Bernadó, Joaquín Arribas, Chow Yin Wong, Puay Hoon Tan, Mihir Gudi, Thomas Choudary Putti, Joohyuk Sohn, Swee Ho Lim, Soo Chin Lee, Yoon Pin Lim
ABSTRACT
Trastuzumab-based chemotherapy has shown remarkable clinical benefits for patients with HER2-positive breast cancer. However, treatment regimens involving trastuzumab had little or no effect for a subset of patients. Preliminary studies revealed WW-binding protein 2 (WBP2), an oncogenic transcription coactivator, to be coamplified with HER2 in 36% of HER2-positive breast cancers. We hypothesize that WBP2 regulates and correlates with the response of HER2-positive breast cancer to trastuzumab. The coexpression of WBP2 and HER2 in breast tumors was validated using IHC. The role and mechanism of WBP2 in regulating breast cancer response to trastuzumab was elucidated using in vitro, patient-derived xenograft and murine xenograft models. A multicenter retrospective study involving 143 patients given neoadjuvant trastuzumab-based chemotherapy was conducted to determine whether WBP2 expression correlates with pathologic complete response (pCR). Elevated expression of WBP2 significantly enhanced breast cancer's response to trastuzumab by augmenting trastuzumab-induced HER2 downregulation and cell-cycle arrest via inhibition of cyclin D expression. High level of WBP2 correlated with better pCR (67.19%) compared with low WBP2 level (26.58%). The highest response was observed in subgroups of patients with high WBP2-expressing tumors also aged below 50 years (77.78%) or were premenopausal in status (73.33%). Retrospectively, WBP2 demonstrated sensitivity of 80% to 81% and specificity of 76.5% to 80% in discriminating between patients showing pCR and non-pCR. WBP2 expression correlates with the response of HER2-positive breast cancer to trastuzumab-based neoadjuvant chemotherapy.