Merck
  • Simultaneous Targeting of PARP1 and RAD52 Triggers Dual Synthetic Lethality in BRCA-Deficient Tumor Cells.
Simultaneous Targeting of PARP1 and RAD52 Triggers Dual Synthetic Lethality in BRCA-Deficient Tumor Cells.
Cell reports (2018-06-14)
Katherine Sullivan-Reed, Elisabeth Bolton-Gillespie, Yashodhara Dasgupta, Samantha Langer, Micheal Siciliano, Margaret Nieborowska-Skorska, Kritika Hanamshet, Elizaveta A Belyaeva, Andrea J Bernhardy, Jaewong Lee, Morgan Moore, Huaqing Zhao, Peter Valent, Ksenia Matlawska-Wasowska, Markus Müschen, Smita Bhatia, Ravi Bhatia, Neil Johnson, Mariusz A Wasik, Alexander V Mazin, Tomasz Skorski
ABSTRACT
PARP inhibitors (PARPis) have been used to induce synthetic lethality in BRCA-deficient tumors in clinical trials with limited success. We hypothesized that RAD52-mediated DNA repair remains active in PARPi-treated BRCA-deficient tumor cells and that targeting RAD52 should enhance the synthetic lethal effect of PARPi. We show that RAD52 inhibitors (RAD52is) attenuated single-strand annealing (SSA) and residual homologous recombination (HR) in BRCA-deficient cells. Simultaneous targeting of PARP1 and RAD52 with inhibitors or dominant-negative mutants caused synergistic accumulation of DSBs and eradication of BRCA-deficient but not BRCA-proficient tumor cells. Remarkably, Parp1-/-;Rad52-/- mice are normal and display prolonged latency of BRCA1-deficient leukemia compared with Parp1-/- and Rad52-/- counterparts. Finally, PARPi+RAD52i exerted synergistic activity against BRCA1-deficient tumors in immunodeficient mice with minimal toxicity to normal cells and tissues. In conclusion, our data indicate that addition of RAD52i will improve therapeutic outcome of BRCA-deficient malignancies treated with PARPi.
Sigma-Aldrich
D-I03, ≥98% (HPLC)