Merck
  • Catalytic properties of the retinal rod outer segment disk ADP-ribosyl cyclase.
Catalytic properties of the retinal rod outer segment disk ADP-ribosyl cyclase.
Visual neuroscience (2011-01-29)
Andrea Fabiano, Isabella Panfoli, Daniela Calzia, Maurizio Bruschi, Silvia Ravera, Angela Bachi, Angela Cattaneo, Alessandro Morelli, Giovanni Candiano
ABSTRACT
Cyclic ADP-ribose (cADPR) is a second messenger modulating intracellular calcium levels. We have previously described a cADPR-dependent calcium signaling pathway in bovine rod outer segments (ROS), where calcium ions play a pivotal role. ROS ADP-ribosyl cyclase (ADPR-cyclase) was localized in the membrane fraction. In the present work, we examined the properties of the disk ADPR-cyclase through the production of cyclic GDP-ribose from the NAD(+) analogue NGD(+). The enzyme displayed an estimated K(m) for NGD(+) of 12.5 ± 0.3 μM, a V(max) of 26.50 ± 0.70 pmol cyclic GDP-ribose synthesized/min/mg, and optimal pH of 6.5. The effect of divalent cations (Zn(2+), Cu(2+), and Ca(2+)) was also tested. Micromolar Zn(2+) and Cu(2+) inhibited the disk ADPR-cyclase activity (half maximal inhibitory concentration, IC50=1.1 and 3.6 μM, respectively). By contrast, Ca(2+) ions had no effect. Interestingly, the properties of the intracellular membrane-associated ROS disk ADPR-cyclase are more similar to those of the ADPR-cyclase found in CD38-deficient mouse brain, than to those of CD38 or CD157. The novel intracellular mammalian ADPR-cyclase would elicit Ca(2+) release from the disks at various rates in response to change in free Ca(2+) concentrations, caused by light versus dark adaptation, in fact there was no difference in disk ADPR-cyclase activity in light or dark conditions. Data suggest that disk ADPR-cyclase may be a potential target of retinal toxicity of Zn(2+) and may shed light to the role of Cu(2+) and Zn(2+) deficiency in retina.
Sigma-Aldrich
Nicotinamide guanine dinucleotide sodium salt, phospodiesterase and ADP-ribosyl cyclase substrate