Merck
  • CO-MP4, a polyethylene glycol-conjugated haemoglobin derivative and carbon monoxide carrier that reduces myocardial infarct size in rats.
CO-MP4, a polyethylene glycol-conjugated haemoglobin derivative and carbon monoxide carrier that reduces myocardial infarct size in rats.
British journal of pharmacology (2008-06-10)
K D Vandegriff, M A Young, J Lohman, A Bellelli, M Samaja, A Malavalli, R M Winslow
ABSTRACT
MP4 (Hemospan) is a Hb-based oxygen therapeutic agent, based on polyethylene-glycol (PEG) conjugation to Hb, undergoing clinical trials as an oxygen carrier. This study describes the functional interaction between MP4 and carbon monoxide (CO), as a CO delivery agent, and the effects of CO-MP4 on myocardial infarct size following ischaemia and reperfusion in rats. Kinetic measurements of CO-MP4 binding were used to evaluate the effects of PEG modification on Hb subunit structure/function and to calculate CO-MP4 equilibrium constants. CO transport by CO-MP4 was shown by ligand (O2/CO) partitioning between MP4 and red blood cell (RBC)-Hb. Pharmacological effects of CO-MP4 were studied on myocardial infarction in rats. CO binding kinetics show primary structural/functional effects on beta chains in MP4, with alpha chains maintaining the ability to undergo tertiary conformational transition. CO confers long-term, room-temperature stability and is able to rapidly re-equilibrate between MP4 and RBCs. In a rat model of myocardial infarct, in contrast to oxy-MP4, CO-MP4 reduced infarct size when administered prior to the induction of ischaemia. MP4 PEGylation chemistry modifies the individual function of Hb subunits, but results in an overall CO equilibrium constant similar to that for unmodified Hb. CO-MP4 is able to deliver CO to the circulation and reduces ischaemia/reperfusion injury in rats, providing the first evidence for this drug as a CO therapeutic agent.
Sigma-Aldrich
Microperoxidase (MP-11) sodium salt, ≥85% (HPLC)