Merck
  • Purification and characterization of cysteine-S-conjugate N-acetyltransferase from pig kidney.
Purification and characterization of cysteine-S-conjugate N-acetyltransferase from pig kidney.
The Biochemical journal (1996-07-01)
A Aigner, M Jäger, R Pasternack, P Weber, D Wienke, S Wolf
ABSTRACT
Microsomal cysteine-S-conjugate N-acetyltransferase catalyses the N-acetylation of various S-substituted cysteines in liver and kidney. We describe here the purification and more detailed characterization of this enzyme catalysing the final reaction of mercapturic acid biosynthesis, and thus playing a crucial role in the detoxicating metabolism of many xenobiotics. The solubilization of cysteine-S-conjugate N-acetyltransferase by deoxy-BIGCHAP [N,N'-bis-(3-D-gluconamidopropyl)deoxycholamide] was the prerequisite for partial purification by means of anion-exchange chromatography. The molecular mass of the enzyme was determined by gel filtration. A polyclonal antiserum was raised against the excised protein band from SDS/PAGE and purified antibodies were used for the complete purification of native cysteine-S-conjugate N-acetyltransferase by immuno-affinity chromatography. A dimeric form of the enzyme was sometimes detected on SDS/PAGE, depending on the degree of purification. For further characterization of cysteine-S-conjugate N-acetyltransferase, the stability of catalytic activity, the pH optimum and K(m) values were determined. The inhibitory effects of various agents were tested, revealing a substantial, yet not complete, loss of cysteine-S-conjugate N-acetyltransferase activity after treatment with cysteine proteinase inhibitors or probenecid under various conditions.