Merck
Mitochondrial myopathy in a german shepherd dog.
Veterinary pathology (2003-09-02)
O Paciello, P Maiolino, G Fatone, S Papparella
ABSTRACT
A 9-month-old male German Shepherd dog was referred for evaluation of progressive exercise intolerance. Clinical examination revealed a stiff, stilted gait and marked atrophy and hypotonia of skeletal muscle. The dog had raised creatine kinase (181 U/liter), lactate dehydrogenase (510 U/liter), and aspartate aminotransferase (123.6 U/liter) levels, suggesting a muscle disease. Histochemical evaluation of muscle biopsies revealed the presence of subsarcolemmal oxidative activity, reduced nicotinamide adenine dinucleotide, and succinate dehydrogenase, and the absence of cytochrome oxidase activity. Ragged red fibers were demonstrated with Gomori trichrome stain. Ultrastructural examination of the muscle confirmed the presence of subsarcolemmal accumulations of mitochondria and morphologically atypical mitochondria.
Sigma-Aldrich
Trichrome Stain LG Solution