Merck
  • Effects of the tumour promoter okadaic acid on intracellular protein phosphorylation and metabolism.
Effects of the tumour promoter okadaic acid on intracellular protein phosphorylation and metabolism.
Nature (1989-01-05)
T A Haystead, A T Sim, D Carling, R C Honnor, Y Tsukitani, P Cohen, D G Hardie
ABSTRACT
Okadaic acid is a polyether derivative of 38-carbon fatty acid, and is implicated as the causative agent of diarrhetic shellfish poisoning. It is a potent tumour promoter that is not an activator of protein kinase C, but is a powerful inhibitor of protein phosphatases-1 and -2A (PP1 and PP2A) in vitro. We report here that okadaic acid rapidly stimulates protein phosphorylation in intact cells, and behaves like a specific protein phosphatase inhibitor in a variety of metabolic processes. Our results indicate that PP1 and PP2A are the dominant protein phosphatases acting on a wide range of phosphoproteins in vivo. We also find that okadaic acid mimics the effect of insulin on glucose transport in adipocytes, which suggests that this process is stimulated by a serine/threonine phosphorylation event.
Sigma-Aldrich
Okadaic Acid (sodium salt)
Sigma-Aldrich
Okadaic Acid, Sodium Salt, Water-soluble analog of Okadaic Acid. Inhibits protein phosphatases 1 and 2A.