Merck
  • Effect of Resveratrol on Reactive Oxygen Species-Induced Cognitive Impairment in Rats with Angiotensin II-Induced Early Alzheimer's Disease †.
Effect of Resveratrol on Reactive Oxygen Species-Induced Cognitive Impairment in Rats with Angiotensin II-Induced Early Alzheimer's Disease †.
Journal of clinical medicine (2018-10-12)
Yu-Te Lin, Yi-Chung Wu, Gwo-Ching Sun, Chiu-Yi Ho, Tzyy-Yue Wong, Ching-Huang Lin, Hsin-Hung Chen, Tung-Chen Yeh, Chia-Jung Li, Ching-Jiunn Tseng, Pei-Wen Cheng
ABSTRACT
Recent studies have indicated that several anti-hypertensive drugs may delay the development and progression of Alzheimer's disease (AD). However, the relationships among AD, hypertension, and oxidative stress remain to be elucidated. Here, we aimed to determine whether reactive oxygen species (ROS) reduction by resveratrol in the brain leads to cognitive impairment reduction in rats with angiotensin II (Ang-II)-induced early AD. Male Wistar Kyoto (WKY) rats with Ang-II-induced AD were treated with losartan or resveratrol for two weeks. Our results show decreased blood pressure, increased hippocampal brain-derived neurotrophic factor (BDNF) level, and decreased nucleus tractus solitarius (NTS) ROS production in the Ang-II groups with losartan (10 mg/kg), or resveratrol (10 mg/kg/day) treatment. Furthermore, losartan inhibition of hippocampal TauT231 phosphorylation activated AktS473 phosphorylation, and significantly abolished Ang-II-induced Aβ precursors, active caspase 3, and glycogen synthase kinase 3β (GSK-3β)Y216 expressions. Consistently, resveratrol showed similar effects compared to losartan. Both losartan and resveratrol restored hippocampal-dependent contextual memory by NADPH oxidase 2 (NOX2) deletion and superoxide dismutase 2 (SOD2) elevation. Our results suggest that both losartan and resveratrol exert neuroprotective effects against memory impairment and hippocampal damage by oxidative stress reduction in early stage AD rat model. These novel findings indicate that resveratrol may represent a pharmacological option similar to losartan for patients with hypertension at risk of AD during old age.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Anti-phospho-GSK3β (Ser9) Antibody, clone 2D3, clone 2D3, Upstate®, from mouse