Merck
  • Multicomponent Mannich reactions with boron enolates derived from diazo esters and 9-BBN.
Multicomponent Mannich reactions with boron enolates derived from diazo esters and 9-BBN.
Organic letters (2011-04-09)
Yi Luan, Scott E Schaus
ABSTRACT
Diazo esters, arylboranes, and carbamoyl imines undergo a 3-component Mannich condensation reaction. Catalyzed by Cu(II) salts, the reaction affords the corresponding isocyanate Mannich product: one that can be easily trapped in situ by other nucleophiles. The Mannich condensation corresponds to an α,α-disubstituted enolate addition to imines. The reaction was rendered asymmetric by using the (-)-phenylmenthol ester in good yield and selectivities.
Sigma-Aldrich
9-Borabicyclo[3.3.1]nonane solution, 0.4 M in hexanes