- Compartmentalised RNA catalysis in membrane-free coacervate protocells.
Compartmentalised RNA catalysis in membrane-free coacervate protocells.
Nature communications (2018-09-09)
Björn Drobot, Juan M Iglesias-Artola, Kristian Le Vay, Viktoria Mayr, Mrityunjoy Kar, Moritz Kreysing, Hannes Mutschler, T-Y Dora Tang
PMID30194374
ABSTRACT
Phase separation of mixtures of oppositely charged polymers provides a simple and direct route to compartmentalisation via complex coacervation, which may have been important for driving primitive reactions as part of the RNA world hypothesis. However, to date, RNA catalysis has not been reconciled with coacervation. Here we demonstrate that RNA catalysis is viable within coacervate microdroplets and further show that these membrane-free droplets can selectively retain longer length RNAs while permitting transfer of lower molecular weight oligonucleotides.