Merck
  • Complex investigation of extraction techniques applied for cyclitols and sugars isolation from different species of Solidago genus.
Complex investigation of extraction techniques applied for cyclitols and sugars isolation from different species of Solidago genus.
Electrophoresis (2018-03-16)
Ileana-Andreea Ratiu, Hossam Al-Suod, Magdalena Ligor, Tomasz Ligor, Viorica Railean-Plugaru, Bogusław Buszewski
ABSTRACT
Cyclitols are phytochemicals naturally occurring in plant material, which attracted an increasing interest due to multiple medicinal attributes, among which the most important are the antidiabetic, antioxidant, and anticancer properties. Due to their valuable properties, sugars are used in the food industry as sweeteners, preservatives, texture modifiers, fermentation substrates, and flavoring and coloring agents. In this study, we report for the first time the quantitative analysis of sugars and cyclitols isolated from Solidago virgaurea L., which was used for the selection of the optimal solvent and extraction technique that can provide the best possible yield. Moreover, the quantities of sugars and cyclitols extracted from two other species, Solidago canadensis and Solidago gigantea, were investigated using the best extraction method and the most appropriate solvent. Comparative analysis of natural plant extracts obtained using five different techniques-maceration, Soxhlet extraction, pressurized liquid extraction, ultrasound-assisted extraction, and supercritical fluid extraction-was performed in order to decide the most suitable, efficient, and economically convenient extraction method. Three different solvents were used. Analysis of samples has been performed by solid-phase extraction for purification and pre-concentration, followed by derivation and GC-MS analysis. Highest efficiency for the total amount of obtained compounds has been reached by PLE, when water was used as a solvent. d-pinitol amount was almost similar for every solvent and for all the extraction techniques involved.
Sigma-Aldrich
allo-Inositol, 97%
Sigma-Aldrich
scyllo-Inositol, ≥98%