Merck
  • Frontline Science: Microbiota reconstitution restores intestinal integrity after cisplatin therapy.
Frontline Science: Microbiota reconstitution restores intestinal integrity after cisplatin therapy.
Journal of leukocyte biology (2018-03-15)
Alfredo Perales-Puchalt, Jairo Perez-Sanz, Kyle K Payne, Nikolaos Svoronos, Michael J Allegrezza, Ricardo A Chaurio, Carmen Anadon, Joseph Calmette, Subir Biswas, Jessica A Mine, Tara Lee Costich, Logan Nickels, Jayamanna Wickramasinghe, Melanie R Rutkowski, Jose R Conejo-Garcia
ABSTRACT
Due to their cytotoxic activities, many anticancer drugs cause extensive damage to the intestinal mucosa and have antibiotic activities. Here, we show that cisplatin induces significant changes in the repertoire of intestinal commensal bacteria that exacerbate mucosal damage. Restoration of the microbiota through fecal-pellet gavage drives healing of cisplatin-induced intestinal damage. Bacterial translocation to the blood stream is correspondingly abrogated, resulting in a significant reduction in systemic inflammation, as evidenced by decreased serum IL-6 and reduced mobilization of granulocytes. Mechanistically, reversal of dysbiosis in response to fecal gavage results in the production of protective mucins and mobilization of CD11b+ myeloid cells to the intestinal mucosa, which promotes angiogenesis. Administration of Ruminococcus gnavus, a bacterial strain selectively depleted by cisplatin treatment, could only partially restore the integrity of the intestinal mucosa and reduce systemic inflammation, without measurable increases in the accumulation of mucin proteins. Together, our results indicate that reconstitution of the full repertoire of intestinal bacteria altered by cisplatin treatment accelerates healing of the intestinal epithelium and ameliorates systemic inflammation. Therefore, fecal microbiota transplant could paradoxically prevent life-threatening bacteremia in cancer patients treated with chemotherapy.
Product Description
Millipore
Yeast Extract, suitable for microbiology
Millipore
Brain Heart Infusion Broth, NutriSelect® Plus, suitable for microbiology