Merck
[Asepsis sowing and tissue culture of Bletilla striata].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2005-02-12)
Songjun Zeng, Xiangli Huang, Zhilin Chen, Jiantong Chen, Jun Duan
ABSTRACT
The asepsis sowing and tissue culture of Bletilla striata were studied. The results indicated that the embyro culture had highest sprouting percent and plantlets percent when their embryos were mature. The optimal medium for the embryo culture was 1/2 MS. Adding 10% coconut juice can promote embyros sprouting and plantlets formation, 1% active carbon can improve plantlets growing. The best medium for the top of plantlet on culture and multiplication was 1/2 MS + 6-BA 0.5 mg/L + NAA 0.2 mg/L. The best medium of rooting was 1/2 MS + NAA 0.5 mg/L, and 10% banana juice can improve rooting of plantlets.
Sigma-Aldrich
Banana powder, suitable for plant cell culture