![Zinc difluoromethanesulfinate 95%](/deepweb/assets/sigmaaldrich/product/structures/334/789/d6c0569c-109d-4304-8b43-7d4d7d8f1e03/640/d6c0569c-109d-4304-8b43-7d4d7d8f1e03.png)
Zinc difluoromethanesulfinate
![Zinc difluoromethanesulfinate 95%](/deepweb/assets/sigmaaldrich/product/structures/334/789/d6c0569c-109d-4304-8b43-7d4d7d8f1e03/640/d6c0569c-109d-4304-8b43-7d4d7d8f1e03.png)
Synonym(s):
Bis(((difluoromethyl)sulfinyl)oxy)zinc, 1,1-difluoro-methanesulfinic acid zinc salt (2:1), Baran difluoromethylation reagent, DFMS
Empirical Formula (Hill Notation):
C2H2F4O4S2Zn
CAS Number:
Molecular Weight:
295.55
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Reaction Type
Reagent Type
Showing 1-1 of 1