Skip to Content
Merck
vibrant-m

Zidovudine Related Compound B

vibrant-m
Synonym(s):
3′-Chloro-3′-deoxythymidine
Empirical Formula (Hill Notation):
C10H13ClN2O4
CAS Number:
Molecular Weight:
260.67

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1