![Zaleplon Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/307/460/e53a60f9-9485-4783-b830-20081a4837a5/640/e53a60f9-9485-4783-b830-20081a4837a5.jpg)
Zaleplon Related Compound A
![Zaleplon Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/307/460/e53a60f9-9485-4783-b830-20081a4837a5/640/e53a60f9-9485-4783-b830-20081a4837a5.jpg)
Synonym(s):
(E)-N-{3-[3-(Dimethylamino)acryloyl]phenyl}-N-ethylacetamide, N-[3-[(2E)-3-(Dimethylamino)-1-oxo-2-propen-1-yl]phenyl]-N-ethylacetamide
Empirical Formula (Hill Notation):
C15H20N2O2
Molecular Weight:
260.33
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2