![TSKgel® SuperMultiporeHZ-N HPLC Column phase polystyrene-divinylbenzene, L × I.D. 15 cm × 4.6 mm, 3 μm particle size](/deepweb/assets/sigmaaldrich/product/images/325/203/b0db4e7a-a5af-43c5-b111-d166c896dd27/640/b0db4e7a-a5af-43c5-b111-d166c896dd27.jpg)
TSKgel® SuperMultiporeHZ-N (3 µm) HPLC Columns
![TSKgel® SuperMultiporeHZ-N HPLC Column phase polystyrene-divinylbenzene, L × I.D. 15 cm × 4.6 mm, 3 μm particle size](/deepweb/assets/sigmaaldrich/product/images/325/203/b0db4e7a-a5af-43c5-b111-d166c896dd27/640/b0db4e7a-a5af-43c5-b111-d166c896dd27.jpg)
Synonym(s):
TSKgel® Size Exclusion (H-Type) HPLC Column
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing