Skip to Content
Merck
TSKgel® GMHXL HPLC Column phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 9 μm particle size

TSKgel® GMHXL (9 µm) HPLC Columns

TSKgel® GMHXL HPLC Column phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 9 μm particle size
Synonym(s):
TSKgel® G1000HXL HPLC Column for Light Scattering Applications

Products

Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1