Skip to Content
Merck
Tropicamide for peak identification European Pharmacopoeia (EP) Reference Standard

Tropicamide for peak identification

Tropicamide for peak identification European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Tropicamide, N-Ethyl-2-phenyl-N-(4-pyridylmethyl)hydracrylamide, Ro 1-7683
Empirical Formula (Hill Notation):
C17H20N2O2
CAS Number:
Molecular Weight:
284.35

Products

Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1