Skip to Content
Merck
(Triethylsilyl)acetylene 97%

(Triethylsilyl)acetylene

(Triethylsilyl)acetylene 97%
Synonym(s):
Ethynyltriethylsilane
Linear Formula:
(C2H5)3SiC≡CH
CAS Number:
Molecular Weight:
140.30

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing