Skip to Content
Merck
Triadimenol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Triadimenol

Triadimenol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
α-tert-Butyl-β-(4-chlorophenoxy)-1H-1,2,4-triazole-1-ethanol
Empirical Formula (Hill Notation):
C14H18ClN3O2
CAS Number:
Molecular Weight:
295.76

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique