
Topiramate impurity E

Synonym(s):
D-(−)-Fructose, D-Levulose, Fruit sugar
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Products
Formula Weight
Manufacturer
Melting Point (°C)
Quality Segment
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing