![Ticarcillin impurity C British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/300/065/ca985014-8a0f-48e3-85fe-33cf9b6b8aca/640/ca985014-8a0f-48e3-85fe-33cf9b6b8aca.png)
Ticarcillin impurity C
![Ticarcillin impurity C British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/300/065/ca985014-8a0f-48e3-85fe-33cf9b6b8aca/640/ca985014-8a0f-48e3-85fe-33cf9b6b8aca.png)
Synonym(s):
3-Thiophenemalonic acid, α-Carboxythiophene-3-acetic acid, (3-Thienyl)malonic acid
Empirical Formula (Hill Notation):
C7H6O4S
CAS Number:
Molecular Weight:
186.19
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing