![Thymosin α1 bovine ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/385/051/7a6094da-ea6a-4bfd-b581-9090a11e9aa5/640/7a6094da-ea6a-4bfd-b581-9090a11e9aa5.png)
Thymosin α1 bovine
![Thymosin α1 bovine ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/385/051/7a6094da-ea6a-4bfd-b581-9090a11e9aa5/640/7a6094da-ea6a-4bfd-b581-9090a11e9aa5.png)
Synonym(s):
Ac-Ser-Asp-Ala-Ala-Val-Asp-Thr-Ser-Ser-Glu-Ile-Thr-Thr-Lys-Asp-Leu-Lys-Glu-Lys-Lys-Glu-Val-Val-Glu-Glu-Ala-Glu-Asn
Empirical Formula (Hill Notation):
C129H215N33O55
CAS Number:
Molecular Weight:
3108.28
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing