Skip to Content
Merck
tert-Amyl methyl ether 97%

tert-Amyl methyl ether

tert-Amyl methyl ether 97%
Synonym(s):
1,1-Dimethylpropyl methyl ether, Methyl 2-methyl-2-butyl ether, Methyl tert-pentyl ether, TAME
Linear Formula:
C2H5C(CH3)2OCH3
CAS Number:
Molecular Weight:
102.17
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing