Skip to Content
Merck
T-2 Toxin from Fusarium sp., powder, ≥98% (HPLC)

T-2 Toxin

T-2 Toxin from Fusarium sp., powder, ≥98% (HPLC)
Synonym(s):
12,13-Epoxytrichothec-9-ene-3,4,8,15-tetrol-4,15-diacetate-8-isovalerate, 4β,15-Diacetoxy-3α-hydroxy-8α-(3-methylbutyryloxy)-12,13-epoxytrichothec-9-ene
Empirical Formula (Hill Notation):
C24H34O9
CAS Number:
Molecular Weight:
466.52
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing