Steviolbioside
Synonym(s):
(4α)-13-[(2-O-β-D-Glucopyranosyl-β-D-glucopyranosyl)oxy]kaur-16-en-18-oic acid, Steviobioside
Empirical Formula (Hill Notation):
C32H50O13
CAS Number:
Molecular Weight:
642.73
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing