
S,S-Dibenzyl trithiocarbonate

Synonym(s):
Carbonic acid, trithio-, dibenzyl ester, Carbonotrithioic acid bis(phenylmethyl) ester, DBTTC, Dibenzyl carbonotrithioate, NSC 33081
Empirical Formula (Hill Notation):
C15H14S3
CAS Number:
Molecular Weight:
290.47
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1