
Sodium triethylborohydride solution

Synonym(s):
Sodium triethylborate, Sodium triethylhydridoborate, Sodium triethylhydroborate
Linear Formula:
(C2H5)3NaBH
CAS Number:
Molecular Weight:
121.99
Products
Brand
Formula Weight
Physical Form
Quality Segment
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing