Serotonin hydrochloride
Synonym(s):
3-(2-Aminoethyl)-5-hydroxyindole hydrochloride, 5-HT, 5-Hydroxytryptamine hydrochloride
Empirical Formula (Hill Notation):
C10H12N2O · HCl
CAS Number:
Molecular Weight:
212.68
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing