![Schizandrin B phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/643/792/df83fba9-4be4-450a-bd1b-572118d2bda7/640/df83fba9-4be4-450a-bd1b-572118d2bda7.png)
Schizandrin B
![Schizandrin B phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/643/792/df83fba9-4be4-450a-bd1b-572118d2bda7/640/df83fba9-4be4-450a-bd1b-572118d2bda7.png)
Synonym(s):
γ-Schizandrin, Schisandrin B, Wuweizisu B
Empirical Formula (Hill Notation):
C23H28O6
CAS Number:
Molecular Weight:
400.46
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing