Rhein 8-glucoside
Synonym(s):
Rhein 8-O-β-D-glucopyranoside, 8-Glucosylrhein, Glucorhein, Rhein 8-glucopyranoside
Empirical Formula (Hill Notation):
C21H18O11
CAS Number:
Molecular Weight:
446.36
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing