Raoultella planticola (formerly Klebsiella aerogenes) NCTC 9528 Lenticule® discs
Products
Brand
Manufacturer
Physical Form
Agency
Quality Segment
Application
Microorganism Suitability
Package Size
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing