![rac-Naproxen Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/214/732/ecd14b80-5944-4471-8e1a-eb40a7699c66/640/ecd14b80-5944-4471-8e1a-eb40a7699c66.png)
rac-Naproxen
![rac-Naproxen Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/214/732/ecd14b80-5944-4471-8e1a-eb40a7699c66/640/ecd14b80-5944-4471-8e1a-eb40a7699c66.png)
Synonym(s):
(±)-Naproxen, 2-(6-Methoxy-2-naphthyl)propionic acid
Linear Formula:
CH3OC10H6CH(CH3)CO2H
CAS Number:
Molecular Weight:
230.26
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing